Would you like to log in again?
Carbide Micro Cutting Tools, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
62094-04-50 62094-01-25 62094-03-50 62094-09-100 62094-03-25 62094-08-50 62094-02-25 62094-21-50 62094-09-25 62094-02-100 62094-01-12 62094-04-12 62094-03-12 62094-13-100 62094-02-12 62094-02-50 62094-03-100 62094-13-25 62094-21-100 62094-21-25 62094-21-12 62094-08-12 62094-04-25 62094-01-100 62094-01-50 62094-04-100 62094-09-12 62094-13-50 62094-08-100 62094-13-12 62094-09-50
103302-160EA 80.26 USD InStock
103302-160 100497-770 100497-750 100497-772 100497-752 100497-774 100497-754 100497-776 100497-756 100497-778 100497-758 102093-942 100501-670 103302-010 100501-674 100501-672 100497-780 100497-760 100501-668 100497-762 100497-764 103302-948 100497-768 100497-748 103302-006 103302-008 100501-666 100501-664 103302-162 103302-662 103302-660
Carbide Micro Cutting Tools, Electron Microscopy Sciences
Cutters
These unique micro cutting tools are made using the latest grinding and polishing technology.
  • Highest quality solid carbide tools
  • Handles are interchangeable in tool handle MT1
Different tip sizes such as 0.12 mm (0.005"), 0.25 mm (0.010"), 0.5 mm (0.020") and 1 mm (0.040") diameter are available.
Ordering information: Each of the tool tips and handle can be ordered separately or in sets. Standard sets come with hardwood instrument cases and include seven tool tips (except micro-graver, 3-sided tool tips) and one handle MT1.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty