Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Stainless Steel Micro Tools, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
62096-08 62096-01 62096-06 62096-02 62096-00 62096-04 62096-05 62096-03 62096-HD 62096-07
100497-804EA 84.21 USD InStock
100497-804 100497-792 100497-796 100501-678 100497-798 100501-676 100497-800 100497-802 100502-474 100503-058
Stainless Steel Micro Tools, Electron Microscopy Sciences
Tool Sets
All tools are forged from the finest quality stainless steel and are precision hand ground, lapped, and polished. These tools are well suited for most applications in the field of microscopic science which demands the ultimate in precision miniature tools. Each tool may be ordered separately or in a set consisting of all 8 tools with mounted tips, one MT handle and one alderwood storage case.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty