Would you like to log in again?
Double Clamp, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
62123-02 62123-01
100497-852EA 47.17 USD InStock
100497-852 100497-854
Double Clamp, Electron Microscopy Sciences
Clamps General Purpose Clamps
A versatile tool which can be adjusted to any position and firmly hold small items of any shape.
Two adjustable arms with alligator jaws in a heavy base. Double Clamp is also available with a 2 ½" diameter magnifier attached.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty