Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Flexible Shaft Machine - General Application Kit, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
62130-10 62131-10 62134-10 62133-20 62134-20 62130-20 62131-20 62133-10
100497-880EA 376.45 USD InStock
100497-880 100497-892 100497-894 100497-884 100497-896 100497-886 100497-898 100497-878
Flexible Shaft Machine - General Application Kit, Electron Microscopy Sciences
Tool Sets
A versatile hang-up flexible shaft machine.
Accessories information: Mounted accessories are on 2.35 mm (3/32") arbors.
Ordering information: Available either 1/10 or 1/8 HP ball bearing motor, variable speed 0 to 18 rpm. Complete kit comes with a hand piece, 1 m (39") Flexible shaft, variable speed foot pedal, and a bracket for hanging. Lifetime warranty on motor under normal use.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty