Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Precision Pin Vise Handle and Diamond Stylus, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
62140-25 62140-02 62140-05 62140-60 62140-50 62140-04 62140-07 62140-06 62140-03 62140-00
100497-936EA 40.42 USD InStock
100497-936 100497-926 102093-944 100497-928 101412-604 100497-930 100497-932 100497-922 100497-934 100497-924
Precision Pin Vise Handle and Diamond Stylus, Electron Microscopy Sciences
Tool Sets
This high precision pin vise handle is machined from the finest material, nonrolling gold-anodized aluminum, which makes it lightweight and versatile.
It features an 8 mm (0.31") hexagonal barrel dimension by 100 mm (4") long (overall length is 115 mm fully assembled) and it has a knurled finger-grip for positive precision control for handling and tool manipulation.
Delivery information: Comes complete with a precision drawbar for positive collets chuck clamping. The vise is supplied with a standard collet chuck opening of 2 mm (0.78") diameter and will hold a selection of our specially designed tools and collets adapter 0.8 mm (0.003") for diamond stylus chucking.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty