Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Hardened Steel Deburring Tool Set, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
62161-SD
100497-998EA 30.78 USD InStock
100497-998
Hardened Steel Deburring Tool Set, Electron Microscopy Sciences
Tool Sets
Made from hardened steel, this tool has three razor sharp cutting edges ranging from a very sharp point to one that is approximately 10 mm in length.
These nickel plated triangular tools are mounted in red and black plastic handles. Tool length is 30 mm by 2.1 mm across (1.2×0.08"). The overall length of the tool is 154 mm (5.7"). Two tools/set.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty