Would you like to log in again?
Adjustable Embedding Mold, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
62361-03 62361-09 62361-07 62361-05 62361-01
100498-048EA 158.56 USD InStock
100498-048 100498-046 100498-044 100498-052 100498-050
Adjustable Embedding Mold, Electron Microscopy Sciences
Moulds Moulds for Histology
These two 'L’s' shape and stainless steel base plate form an adjustable embedding mold, suitable for embedding various size specimens with a minimum of waste material.
Available in five different sizes.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty