Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Embedding Blocks, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
62363-02 62363-01 62363-04 62363-03
100498-060EA 184.6 USD InStock
100498-060 100498-058 100498-056 100498-054
Embedding Blocks, Electron Microscopy Sciences
Embedding Cassettes Embedding Rings
Embedding blocks are precisely machined from hard phenolic resin
It is suitable for cellodin or paraffin sections. The faces of the embedding blocks are cut very deeply providing a firm hold of the embedded specimen.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty