Would you like to log in again?
Micro Spoon and Spatula, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
62410-01
100498-062EA 12.59 USD InStock
100498-062
Micro Spoon and Spatula, Electron Microscopy Sciences
Spatulas
A micro spoon and spatula made from nickel stainless steel and coated with a glazed finish resin on the handle.
  • Overall length: 165 mm (6 ¹/₂")
  • Spoon L×W: 25×6.35 mm (1×¹/₄")
The wire is 0.103" (2.62 mm) in Ø.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty