Would you like to log in again?
Inoculating Loops, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
62438-03 62437-03 62434-03 62434-05 62438-02 62433-05 62433-03 62436-05 62433-02 62436-03 62434-02 62437-02 62437-05
100498-108EA 267.22 USD InStock
100498-108 100498-106 100498-104 100498-102 100498-100 100498-112 100498-110 100498-088 100498-086 100498-084 100498-092 100498-082 100498-090
Inoculating Loops, Electron Microscopy Sciences
Spreaders Inoculating Loops and Needles
Our high quality inoculating loops are made from platinum wire. The 15% iridium adds necessary rigidity permitting thinner wire for delicate work. The loop section is welded and smoothly finished to form a “perfect circle”.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty