Would you like to log in again?
Instrument Trays With Cover, Natural polypropylene, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
63312-03 63312-01 63312-02
100498-406EA 83.71 USD InStock
100498-406 100498-404 100498-408
Instrument Trays With Cover, Natural polypropylene, Electron Microscopy Sciences
Trays/Dishes/Pots Instrument Trays/Dishes
The cover protects the instruments from contamination.
  • Natural polypropylene
  • They are easily cleaned and sterilized
Steam autoclavable at 121 °C (250 °F), smooth with rounded corners.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty