Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Colormark ™; Plus Glass Slide, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
Colormark®
63400-41 63400-20 63400-50 63400-61 63400-10 63400-40 63400-51 63400-21 63400-11 63400-60 63400-31 63400-30
100491-382GR 154.77 USD InStock
100491-382 100491-380 100498-458 100491-386 100498-456 100491-374 100491-384 100491-378 100491-376 100498-462 100502-512 100498-460
Colormark ™; Plus Glass Slide, Electron Microscopy Sciences
Slides Microscope Slides
Specimen slide identification is vital automated etched slide labeling instruments ensure specimen slide integrity but are difficult to read on ordinary slides.
To clearly read diamond scribed slide labels against any surface, you need the only slide designed to be used with your automated slide labeling instrument.
You need COLORMARK slides.These brightly printed slides greatly improve etched labeling visibility and offer all the advantages of Superfrost slides. A bright contrasting color has been printed on the back of the Superfrost end so that patient or specimen information etched through the painted end on the front is clearly visible. Colormark Plus slides are adhesive coated therefore ensuring the adhesion of the section to the slide during staining procedures.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty