Would you like to log in again?
Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Saturday, May 25. 
Stirring Bar Retrievers, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
64208-12 64208-13 64208-18
100498-648EA 34.61 USD InStock
100498-648 100498-650 100502-532
Stirring Bar Retrievers, Electron Microscopy Sciences
Stirrers Stirring Tools
Polypropylene Stirring Bar Retrievers
They are ideal for use with all stirring bars. The entire retriever is totally encased in a chemically resistant polypropylene, with a handy ring for hanging when not in use. PTFE Stirring Bar Retriever: A permanent powerful magnet encapsulated in PTFE and supplied with flexible PTFE cord.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty