Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Automatic Pipette Washing Set, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
64377-05 64377-15 64377-25
100498-746EA 2534.76 USD InStock
100498-746 100498-752 100502-542
Automatic Pipette Washing Set, Electron Microscopy Sciences
Pipettes Pipette Washers
Solve the problem of washing pipettes quickly, safely, and with little effort. To use allow dirty pipettes to soak in the pipette basket in the pipette jar and when ready the basket is transferred to the washer where a constant flow of tap water rinses the pipettes clean.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty