Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Pipettor Holder, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
64378-04 64378-06 64378-6B
100498-758EA 149.19 USD InStock
100498-758 100498-756 100498-760
Pipettor Holder, Electron Microscopy Sciences
Pipette Stands
Made from High Density Polyethylene. Accommodate most major brands of pipettors.
  • Resistant to most solvents and is easy to clean
  • Supplied with non-skid feet
  • Available in 4-place and 6-place versions
Also Available in Black HDPE/UV, perfect for use in DNA Workstations, where U.V. anti-germicidal amp are used.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty