Would you like to log in again?
Scratch Brushes with Metal Ferrules, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
65582-01 65582-07 65582-05 65582-03
103302-488EA 17.02 USD InStock
103302-488 100498-896 103302-490 100498-890
Scratch Brushes with Metal Ferrules, Electron Microscopy Sciences
Brushes
These scratch brushes with pen shaped handles are used for fine deburring, polishing, as well as rust removal and the cleaning and removal of deposited wax.
  • Available with steel, nylon, fiberglass or brass
  • With metal ferrules for improved durability
  • Overall length is 41/4" and the brush is 1/8" wide
Packaging: Refills are available and are packaged 12/box.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty