Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Whipple Grid Graticules, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
68009-27 68009-25 68009-24.5 68009-20.4 68009-23 68009-16 68009-26 68009-19 68009-21
100499-128EA 215.86 USD InStock
100499-128 103305-428 103305-438 100499-130 103305-434 100499-132 103305-436 103305-430 103305-432
Whipple Grid Graticules, Electron Microscopy Sciences
Graticules Reticles and Grids
Whipple grid graticules are used for drinking water particle analysis and for other particle counting.
  • Whipple grid 100 squares in 7 mm area
  • Surface chrome image
Grid showed ratio of full square to smallest is 50:1 and area is 2500:1.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty