Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Indexed Grids, Electron Microscopy Sciences

Supplier: Electron Microscopy Sciences
68018-21A 68016-26 68011-19A 68016-25 68016-24.5 68016-23 67981-24 68016-20.4 68018-16A 68016-19 67981-26 67981-23 68011-16A 68016-16 67981-21 67981-50 68016-21 68016-27 68011-21A 68018-19A
102096-052EA 182.98 USD InStock
102096-052 100499-190 103303-610 102096-050 100499-180 103303-602 102096-112 102096-056 103303-612 102096-110 103303-604 102096-054 100499-178 100499-146 103303-606 100499-198 103303-608 100499-176 102096-058 100499-194
Indexed Grids, Electron Microscopy Sciences
Graticules Reticles and Grids
Useful for particle counting and in area of specimen determinations.

  • NE10A: numbered grid 5×5 mm, 0.5 mm pitch
  • NE11A: numbered grid 10×10 mm, 1.0 mm pitch
  • NE34A: numbered grid 1×1 mm, 0.1 mm pitch
  • NE71: index pattern 20×20 grid of 0.5 mm squares

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty