Would you like to log in again?
Weibel 3 Graticles, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
68023-16 68023-21 68023-19
100499-226EA 424.56 USD InStock
100499-226 100499-228 100499-224
Weibel 3 Graticles, Electron Microscopy Sciences
Graticules Reticles and Grids
Mertz for stereology. Surface chrome image.
Reference: E.R. Weibel, G.S. Kishtler & W.F. Scherle. 1986. J. Cell Biology.30, 23.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty