Would you like to log in again?
Stage Graticules and Micrometers for Transmitted Light, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
68039-16 68043-21 68042-08 68039-11 68040-01 68044-12 68039-22 68039-05 68041-48 68039-04 68043-20 68039-02
100499-314EA 393.32 USD InStock
100499-314 100499-304 100499-316 100499-306 100499-310 100499-300 100499-312 100499-324 100499-302 100499-320 100499-318 100499-308
Stage Graticules and Micrometers for Transmitted Light, Electron Microscopy Sciences
Graticules Reticles and Grids
The stage graticules are for the routine calibration of eyepiece patterns when alternating between objectives or using the graticule in different microscopes.
  • Robust construction
  • Ideal for student use and for instructional purposes
The scale or grid is centered on a glass disc mounted in a black anodized aluminum slide 75×24×2 mm thick.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty