Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
LifterSlip™ Premium Printed Coverglass, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
72186-40 72184-40 72185-24 72184-44 72186-42 72184-30 72186-54 72186-25 72186-44 72186-35 72186-36 72184-50
100499-622EA 143.99 USD InStock
100499-622 100499-612 100499-624 100499-602 100499-630 100499-620 100499-608 100499-604 100499-626 100499-606 100499-628 100499-618
LifterSlip™ Premium Printed Coverglass, Electron Microscopy Sciences
Cover Glasses
LifterSlip™ Premium Printed Coverglass offers a simple solution to the problem of hybridization non-uniformity. An innovative raised edge design provides separation and allows for even dispersal of hybridization solution between the DNA chip and coverslip.
  • 1mm thick
  • Uniform disposal of hybridization solution
  • Reduced hybridization fluid volume variations
  • Superior flatness - substrate does not flex or bow
  • Precision cut edges
  • Superior surface quality
  • Consistent flatness
LifterSlips™ allow for increased data quality by eliminating gradients caused by floating standard coverslips on hybridization solutions. The printed bars allow for easy injection of hybridization solution under the pre-positioned coverslips. Capillary attraction ensures that LifterSlip™ stays in place. Depending on size and design configuration LifterSlip™ can hold a range of volumes. LifterSlip™ is #2 thickness (0.20-0.22 mm) and the bar height is 0.04-0.06 mm.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty