Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Dip Miser, For Slide Coating, 10 ml Capacity, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
70520 70510 70530
100491-618EA 181.45 USD InStock
100491-618 100499-696 100491-620
Dip Miser, For Slide Coating, 10 ml Capacity, Electron Microscopy Sciences
Stains Microscopy and staining reagents
Slide coating cup with stand and cleaning brush. Glass cup is Pyrex and is designed to conserve expensive autoradiographic emulsion when coating glass slides. Needs only 1/4 the volume of solution previously required. Single piece holder permits immersion in a water bath and provides a stable darkroom stand to prevent spills, and faster temperature adjustment of the emulsion.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty