Would you like to log in again?
Dumont Tweezers Style 5, Dumont Switzerland, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
72700-D 72700-DZ 72705-12 72701-12 0103-5-TF 72705-01 72877-DZ 72706-12 72701-01 72877-D 72706-01 0103-5-PO
100501-806PR 63.53 USD InStock
100501-806 100493-834 100499-778 100493-836 100499-762 102091-536 76375-932 100499-780 102098-064 102098-062 100493-852 100499-826
Dumont Tweezers Style 5, Dumont Switzerland, Electron Microscopy Sciences
Forceps
Dumont positive action Style 5 tweezers have super thin tips.
  • Hand made of the highest quality steels and alloys
  • High precision Swiss manufacturing
  • Perfectly finished to suit your application
Dumoxel® is composed of C, Cr, Ni, Mo and Cu and is the most popular alloy among our customers. This alloy was developed and patented by Dumont to offer the best in resistance to corrosion thanks to its high concentration of molybdenum and chromium. Dumoxel® also guarantees an excellent resistance to sulphuric environments, hydrochloric acid, as well as to all other mineral and organic acids. Dumoxel® is 95% antimagnetic, resistant up to temperatures of around 400 °C (DIN 50 914) and is suitable for autoclave sterilization at 270 °C.
Dumostar®, Dumont's patented alloy, is composed of C, Cr, Mo, Mn, Co, Ni and Si. Dumostar is more elastic and more resistant to corrosion than the best stainless steels. It is also perfectly compatible with human tissues and is resistant to mineral and organic acids as well to salt. Although slightly more expensive than other alloys, Dumostar is 100% antimagnetic and is resistant to sterilization temperatures of up to 500 °C.This is the most cost effective and appropriate alloy for laboratory use.
Inox 08 is a medical stainless steel composed of C, Mn, Cr, Mo and V that provides an excellent resistance to corrosion and a good resistance to salt. Although not as hard as Carbon steels, Inox 08 offers excellent resistance to corrosion. This magnetic alloy supports temperatures of around 400 °C (DIN 50914) and is suitable for autoclave sterilization at 180 °C.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty