Would you like to log in again?
Tweezers Style 7, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
72802-D 0304-7-PO 72802-DZ 0303-7-PO
100488-312PR 56.19 USD InStock
100488-312 102091-658 100499-806 102091-700
Tweezers Style 7, Electron Microscopy Sciences
Forceps
Dumont positive action style 7 tweezers have thin curved tips.
  • Handmade of the highest quality steels and alloys
  • High precision Swiss manufacturing
  • Perfectly finished to suit your application
Dumoxel® is composed of C, Cr, Ni, Mo and Cu and is the most popular alloy among our customers. This alloy was developed and patented by Dumont to offer the best in resistance to corrosion thanks to its high concentration of molybdenum and chromium. Dumoxel® also guarantees an excellent resistance to sulphuric environments, hydrochloric acid, as well as to all other mineral and organic acids. Dumoxel® is 95% antimagnetic, resistant up to temperatures of around 400 °C (DIN 50 914) and is suitable for autoclave sterilization at 270 °C.
Inox 02 is a standard magnetic stainless steel composed of C, Mn, Cr and Si. This alloy is not as hard as carbon steels because it contains Chromium. Lnox 02 will resist temperatures of around 400 °C (DIN 50 914) but cannot be sterilized.
Antimagnetic is composed of C, Cr, Ni and Mo. It offers a good resistance to corrosion thanks to its high concentration of molybdenum and is 80% antimagnetic but is not as hard as Inox. Antimagnetic is resistant up to temperatures of around 400 °C (DIN 50 914) and is suitable for autoclave sterilization at 270 °C.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty