Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Rubis Standard SA Steel Tweezers, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
78130-01 78150-12 78139-01 78150-01 78140-12 78141-01 78135-12 78154-12 78135-01 78152-12 78141-12 78140-01 78131-12 78138-01 78152-01 78138-12 78130-12 78154-01 78131-01 78139-12
100502-570DZ 354.11 USD InStock
100502-570 100499-846 100502-572 100499-858 100499-848 100501-494 100494-622 100499-854 100494-630 100499-856 100494-620 100499-860 100499-850 100499-852 100494-618 100494-616 100494-628 100494-626 100501-496 100494-624
Rubis Standard SA Steel Tweezers, Electron Microscopy Sciences
Forceps
The material used in these tweezers is SA steel, which is acid-proof, stainless steel and 100% antimagnetic.
With more than fifty years of experience, RUBIS tweezers represents authentic Swiss quality
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty