The anterior printing area is a 45° angle which suits most cassette labeling instruments
Made from acetal copolymer – This newly patented cassette, allows you to load the Micro-Writer® and Shur/Mark® Cassette Labeling Instruments in one simple operation. Each convenient stack contains 50 unique cassettes. No more manual insertion one-by-one. Save time and money. Two types of cassettes are offered in order to suit your needs: Slotted tissue cassettes and meshed biopsy cassettes.
The anterior printing area is a 45° angle which suits most cassette labeling instruments. Lids are available as one-piece disposable plastic. It can be opened and closed as often as necessary and it always relocks securely without danger of specimen loss. Cassettes are packaged in stacks of 50. Each case contains 2 boxes of 20 stacks. Order cassette bases and cover separately.
Lids are available as one-piece disposable plastic. It can be opened and closed as often as necessary and it always relocks securely without danger of specimen loss. Cassettes are packaged in stacks of 50. Each case contains 2 boxes of 20 stacks. Order cassette bases and cover separately.