Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-VIM Mouse Monoclonal Antibody [clone: 4A4]

Supplier: Enzo Life Sciences
KAM-CC249-E
10050-128EA 436 USD InStock
10050-128
Anti-VIM Mouse Monoclonal Antibody [clone: 4A4]
Antibodies
Vimentin is one of four type II intermediate filament (IF) proteins; Desmin, Glial fibrillary acidic protein (GFAP) and Peripherin being the others. Vimentin is the most widely distributed of all IF proteins. It is typically expressed in leukocytes, blood vessel endothelial cells, and mesenchymal cells such as fibroblasts. Vimentin filaments help support cellular membranes. Vimentin networks also may help keep the nucleus and other organelles in a defined place within the cell.

Type: Primary
Antigen: VIM
Clonality: Monoclonal
Clone: 4A4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty