Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-GRP78 Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
SPA-769-0050
10050-180EA 186 USD InStock
10050-180
Anti-GRP78 Rabbit Polyclonal Antibody
Antibodies
Grp78 (Glucose Regulated 78kDa Protein), is also known as BiP or Immunoglobulin heavy chain binding protein. It is a stressresponse protein which is induced by agents or conditions that adversely affect endoplasmic reticulum (ER) function. This protein is essential for the proper glycosylation, folding, and assembly of many membrane bound and secreted proteins as well as for maintenance of cell homeostasis and the prevention of apoptosis.

Type: Primary
Antigen: Grp78
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-Terminus
Host: Rabbit
Isotype:
Reactivity: Mouse, Rat, Xenopus, Rabbit, Monkey, Hamster, Fungus, Bovine

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty