Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ADGRE1 Rat Monoclonal Antibody (Biotin) [clone: BM8.1]

Supplier: Tonbo Biosciences
30-4801-U100 30-4801-U500 30-4801-U025
10050-774EA 215 USD InStock
10050-774 10050-772 10050-770
Anti-ADGRE1 Rat Monoclonal Antibody (Biotin) [clone: BM8.1]
Antibodies
The BM8.1 antibody is specific for mouse F4/80 antigen, a 125 kDa transmembrane protein widely expressed by members of the mononuclear phagocyte system and considered to be a key marker for mature macrophage cells. F4/80 is differentially expressed during myeloid cell development, and may be regulated by certain cytokines within the tissue microenvironment. Other cell types shown to express this antigen include Langerhans cells, Kupffer cells and dendritic cell subsets. BM8.1 is widely used together with antibodies to CD115 (c-fms), CD11b and CD11c to identify myeloid / macrophage cells by flow cytometry.

Type: Primary
Antigen: ADGRE1
Clonality: Monoclonal
Clone: BM8.1
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: IgG2b kappa
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty