Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Histology Cassettes, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
70070-GR 70071-O 70070-O 70070-P 70070-A 70071-B 70070-Y 70070-G 70071-GR 70071-G 70071-T 70071-L 70070-T 70071-Y 70071-PE 70070-B 70071-P 70070-W 70070-L 70070-PE 70071-A 70071-W
100500-024CS 376.72 USD InStock
100500-024 100500-002 100500-014 100500-026 100500-004 100500-016 100500-006 100500-028 102096-572 100500-018 100500-008 100500-030 100500-020 100500-010 100500-000 100500-022 100499-996 100499-998 100499-988 100499-992 100499-994 100499-990
Histology Cassettes, Electron Microscopy Sciences
Embedding Cassettes Tissue Processing Cassettes
Lids are packed separately
Made from Acetyl Polymer, which is a special high density polymer that keeps specimens safely submerged in solution. These cassettes are totally resistant to most histological solvents and their slots offer maximum flow through for good fluid exchange and proper drainage. The lids either come attached to the cassette or are packaged separately. The lids can be opened and closed as often as necessary and they always relock securely without danger of specimen loss. Large labeling areas are located on three sides of the cassette for your convenience. Each slot measures: 1 x 5 mm, total 128 slots per cassette.
Lids are packed separately. Each case contains two dispenser boxes of 500 cassettes and one dispenser box of 1000 lids.
Lids are placed on cassettes. Each case contains three dispenser boxes of 500 cassettes with lids already locked in place.
Each case contains two dispenser boxes of 500 cassettes and one dispenser box of 1000 lids.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty