Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Biopsy Processing Embedding Cassettes, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
70073-B 70072-B 70072-L 70073-A 70073-L 70073-P 70073-Y 70072-T 70073-GR 70073-PE 70072-GR 70073-G 70072-PE 70072-W 70072-Y 70072-O 70073-T 70072-A 70072-G 70073-O 70073-W 70072-P
100500-056CS 385.47 USD InStock
100500-056 100500-034 100500-068 100500-046 100500-058 100500-036 100500-048 100500-038 100500-070 100500-060 100500-050 100500-072 100500-040 100500-062 100500-074 100500-052 100500-042 100500-064 100500-054 100500-032 100500-066 100500-044
Biopsy Processing Embedding Cassettes, Electron Microscopy Sciences
Embedding Cassettes Biopsy Processing Cassettes
Biopsy processing cassettes are similar to the slotted cassettes but have a 1 mm square mesh for small biopsies. Made from Acetyl Polymer, which is a special high density polymer that keeps specimens safely submerged in solution, these cassettes are totally resistant to most histological solvents and their slots offer maximum flow through for good fluid exchange and proper drainage.
  • Lids either come attached to the cassette or are packaged separately
  • Lids can be opened and closed as often as necessary and always relock securely without specimen loss
  • Large labeling areas are located on three sides of the cassette for your convenience
  • Each slot measures: 1 x 5 mm, total 128 slots per cassette
Packaging: Cassettes with lids are placed on cassettes, each case contains 3 dispenser boxes of 500 cassettes with lids already locked in place. Cassettes with lids packed separately, each case contains two dispenser boxes of 500 cassettes and Lids.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty