Would you like to log in again?
Glass Coplin Staining Jar, Screw Cap, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
70315
100500-230CS 222 USD InStock
100500-230
Glass Coplin Staining Jar, Screw Cap, Electron Microscopy Sciences
Jars Staining Jars
Staining Jar holds 5 single 1x3" slides vertically, or 10 slides back-to-back
Slides extend above the opening, allowing for the manipulation of them without the use of forceps. A screw cap has a metal foil liner that withstands stains and organic solvents and it prevents evaporation.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty