Would you like to log in again?
Glass Coplin Staining Jar, Screw Cap, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
70317
100500-232CS 261.25 USD InStock
100500-232
Glass Coplin Staining Jar, Screw Cap, Electron Microscopy Sciences
Jars Staining Jars
Staining Jar holds 5 single 1 x 3" slides vertically, or 10 slides back-to-back.
Slides stay within the jar so that it can be capped during staining. Complete with a screw cap and a metal foil liner.
Overall measurement: 4 3/8 x 2 3/4 x 2 3/8.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty