Would you like to log in again?
High-Density Polyethylene Staining Jar, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
70319
100500-234CS 68.41 USD InStock
100500-234
High-Density Polyethylene Staining Jar, Electron Microscopy Sciences
Jars Staining Jars
An unbreakable, high-density polyethylene staining jar with internal grooves and a shallow screw cap.
The jar holds 5 single 1 x 3" slides or 10 back-to-back slides vertically.
Overall measurement H x Dia: 4 1/2 x 2 1/2".
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty