Would you like to log in again?
Dust-Off XL Model (DPSXL), Electron Microscopy Sciences
Supplier: Dust-Off
70837 70838
100500-238CS 297.47 USD InStock
100500-238 100502-796
Dust-Off XL Model (DPSXL), Electron Microscopy Sciences
Sprays
Pressurized duster removes dust and dirt with pure moisture free blasts. There is more product at less cost per ounce. Extension tube is included with the can. Its new concentrated formula will give you 57% more blasts per ounce, 17% more blasts per can. Safe for the environment. Contains: Formula S (Difluoroethane) - 100% safe for the ozone.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty