Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Formalin 10% in aqueous solution low odor, buffered

Supplier: Electron Microscopy Sciences
Warning
Synonyms: MethylaldehydeFormolParaformaldehyde aqueous solutionFormaldehyde

15741-04 15741-01
100500-246CS 442.31 USD InStock
100500-246 100496-506
Formalin 10% in aqueous solution low odor, buffered
Formalin
10% formalin in Phosphate buffer according to the A.F.I.P. Manual of Histology.

  • Low odor

This solution allows for long-term storage of tissue specimens.


Formula: CH₂O
MW: 30.03 g/mol
Boiling Pt: 100 °C (212 °F)
Density: 0.83 g/cm³ (6.926 lbs/gal.)
Storage Temperature: Ambient
MDL Number: MFCD00003274
CAS Number: 50-00-0
UN: 3334
ADR: 9,
https://us-legacy.vwr.com/stibo/web/std.lang.all/48/24/28984824.jpg
Order Now
Specification Test Results
pH at 25 °C 6.9 - 7.1

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty