Synonyms:
Methylaldehyde
, Formol
, Paraformaldehyde aqueous solution
, Formaldehyde
15742-30
15742-60
15742-10
15742-20
100500-248CS
98.58
USD
InStock
100500-248
100502-586
100502-588
100503-120
Formalin 10%, TissuePrep, buffered
Formalin
Ready-to-use, prefilled with 10% formalin.
Prefilled with 10% formalin Cat# 15740. Ready-to-use in pre-filled cups. Labeled in accordance to OSHA requirements, which gives you immediate information regarding handling and first aid procedures. Pre-measured in wide-mouth, leak-proof, opaque polypropylene containers which limits your exposure to formaldehyde and saves you time.