Would you like to log in again?
Hollande's Fixative, Electron Microscopy Science
Supplier: Electron Microscopy Sciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
64120-04 64120-01 64120-20 64120-30
100498-614EA 73.08 USD InStock
100498-614 100500-432 100502-582 100498-616
Hollande's Fixative, Electron Microscopy Science
Stains Microscopy and staining reagents
Hollande's fixative is a modification of Bouin's solution. Used as a fixative for biopsy specimens of the gastrointestinal tract.
  • Stable and will decalcify small bone specimens
  • Prefilled ready-to-use
A modification of Bouin's solution. It is stable and will decalcify small bone specimens. Tissue that is fixed with Hollande's can be stained successfully with most stains, and the cupric acetate in the solution stabilizes red blood cell membranes and cosinophil and endocrine cell granules so that less lysis occurs than with Bouin's solution. Hollande's is widely used as a fixative for biopsy specimens of the gastrointestinal tract.
Thorough washing of the fixative prior to placing the specimen in a phosphate buffered formalin solution is necessary because the salts present in the solution will form an insoluble phosphate precipitate. Packaged in leak-proof histo-containers; prefilled ready-to-use.
Packaging: Packaged in leak-proof histo-containers.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty