Would you like to log in again?
Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Saturday, May 25. 
Nunc™ 384 Well® Optical Bottom Plates – Polymer Base, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
64810-50 64810-54 64810-40 64810-25 64811-20 64810-52 64810-35
100500-486CS 974.99 USD InStock
100500-486 100500-498 102095-728 100500-490 100500-492 102095-730 102095-732
Nunc™ 384 Well® Optical Bottom Plates – Polymer Base, Electron Microscopy Sciences
Microplates Cell Culture Plates
The NUNC™ Coverglass Optical Bottom 96- and 384- Well Plates have an optically clear polystyrene plate bottom
This clear base permits microscopic viewing and reading of well The plate’s upper structure is offered in black (for fluorometric detection).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty