Would you like to log in again?
Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Saturday, May 25. 
Process Embedding Cassettes, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
70025-B 70025-GR 70025-W 70025-PE 70025-T 70025-P 70025-Y 70025-O 70025-10 70025-11 70025-L 70025-G 70025-C 70025-A
100500-540CS 76.14 USD InStock
100500-540 100500-552 100502-040 100500-542 100500-554 100500-532 100500-544 100500-534 100500-546 100500-536 100500-550 100490-726 100500-548 100500-538
Process Embedding Cassettes, Electron Microscopy Sciences
Embedding Cassettes Biopsy Processing Cassettes
Made from Acetal copolymer – Disposable plastic tissue cassettes are suitable for holding and identifying tissue samples in processing, embedding, and sectioning procedures. The cassettes fit securely in microtome chuck adapters. They are molded from a high-density polymer that is totally resistant to histological solvents. These cassettes are designed to accept standard metal lids and will also fit either the standard stainless steel base molds or the disposable plastic ones. Only Clear cassettes are made of polypropylene. All other cassettes are made of Acetal Polymer. Each case contains 3 dispenser boxes of 500 cassettes. Stainless Steel Lids order separately.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty