Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Lab-Tek® Chamber Slide™ System, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
70383 70417 70416 70390 70386 70406 70420 70415 70423 70391 70384 70421 70397 70418 70419 70393 70403 70410 70411 70387 70414 70394 70404 70400 70380 70401 70413 70407 70381 70396 70422
100500-596BX 340.79 USD InStock
100500-596 100500-598 100500-610 100500-612 100501-174 100488-388 100499-898 100499-894 100499-896 100500-614 100500-616 100488-382 100488-384 100500-618 100488-386 100500-620 100500-600 100501-768 100500-602 100499-902 100499-900 100488-390 100501-766 100500-604 100501-722 100501-764 100488-392 100500-606 100488-394 100500-608 100488-396
Lab-Tek® Chamber Slide™ System, Electron Microscopy Sciences
Slides Chamber Slides
Permanox® is a special molding resin, which meets the growing requirements of most anchorage dependent cells; also excellent chemical resistance allows for cell fixation and the staining of organic solvents for EM and other techniques. Low oxygen permeation makes Permanox® ideal for anoxic cell growth.
  • Developed for microscopy of in situ cell culture
  • Cells grow on a standard microscope slide
  • Media chamber can be removed when culturingis complete
  • Useful for viral and mycoplasma testing, chromosome studies toxicity tests and immunocytology
  • Permanox® plastic and glass do not interfere with fluorescent labels
The chamber slides combine a media-containing chambered super-structure with a standard soda lime glass slide or Permanox® plastic slide, sealing the two together with a biologically inert silicone gasket. After cells are grown, the chamber and gasket are easily removed. The slide may then be fixed, stained, cover slipped, and filed with the usual convenience. These slides, once fixed and stained, can be embedded using our Slide Duplicating Mold (Cat. 70170). Lab-Tek® Chamber Slide™ System all come packed in a sterile sealed tray, 8 units per tray, 2 trays per pack and 12 trays per case.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty