Would you like to log in again?
Wooden Sticks; Tongue Depressors, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
72322 72320
100500-652BX 36.32 USD InStock
100500-652 100488-474
Wooden Sticks; Tongue Depressors, Electron Microscopy Sciences
Tongue Depressors
These sticks are made from close-grained northern white birch because of its ability to provide a smooth and polished surface.
Used for mixing, stirring, or scraping epoxy resins.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty