Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Compartment - Micro Processing/Embedding Cassettes, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
70078-L 70078-T 70078-W 70078-GR 70078-Y 70078-PE 70078-G 70078-B 70078-O 70078-P 70078-A
100500-750BX 263.79 USD InStock
100500-750 100500-740 100500-752 100500-742 100500-732 100500-744 100500-734 100500-746 100500-736 100500-748 100500-738
Compartment - Micro Processing/Embedding Cassettes, Electron Microscopy Sciences
Embedding Cassettes Biopsy Processing Cassettes
Made from Acetal Polymer. These biopsy cassettes can hold up to six tissue specimens, each one placed in it’s own compartment, numbered from 1 to 6 and measuring 7 x 12 x 5 mm (1/4 x 7/16 x 3/16"). Cover and base have over 3000 0.36 mm square openings. Cassettes keep specimen safely submerged in solvent and are totally resistant to the most histological solvents. The efficient flow-through slots maximize fluid exchange and insure proper drainage. The one-piece integral lid eliminates the need for separate steel lids. Large labeling areas are located on three sides of the cassette. The anterior writing area is at a 45° angle, making the cassette, to some degree, able to be used with certain types of cassette labeling instruments. Each case contains 4 dispenser boxes of 250 cassettes with covers assembled.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty