Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Micro Cover Glasses, Square and Rectangular Sizes, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
72200-10 72210-20 72200-30 72200-21 72210-21 72210-11 72200-40 72210-10 72200-41 72204-03 72204-30 72198-10 72204-10 72204-20 72200-31 72198-11 72204-02 72204-04 72204-01 72200-20 72204-40
100500-820BX 18.93 USD InStock
100500-820 100500-810 100500-822 100500-800 100502-850 100502-840 100493-672 100502-852 100502-842 100502-848 100502-838 100502-854 100502-844 100502-846 100500-812 100500-814 100500-804 100500-816 100500-806 100500-818 100500-808
Micro Cover Glasses, Square and Rectangular Sizes, Electron Microscopy Sciences
Cover Glasses
The Micro Cover Glasses are manufactured from clear white, borosilicate glass. They offer super clarity, are perfectly flat, and corrosion resistant.
  • Uniform in size and thickness
  • Square slides are 22 x 22 mm
  • Rectangular slides are 22 mm wide and 30, 40 or 50 mm long
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty