Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Chemicals for MohrPro, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
74962-01 74963-01 74966-01 74965-01 74962-10 74961-01 74966-03 74961-10 74964-01 74965-03
100494-314EA 24.32 USD InStock
100494-314 100501-164 100494-312 100500-840 100500-842 100500-844 100500-838 100503-158 100494-318 100494-316
Chemicals for MohrPro, Electron Microscopy Sciences
Film Processing Supplies
All chemicals are concentrated. One quart (0.95L) of concentrate mixed with water will make one gallon of working solution. The MohrPro is also available in a 22” model. MOHRPRO 20 model is a 56 cm (22") wide processor with a “Deep Tank” roller transport design capable of processing 240 - 8x10 RA color prints 54.7 cm (18”/minutes) or 480 - 8x10 B&W RC prints (36”/minute) dry to dry per hour.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty