Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.

Thermanox® Cover Slips, Electron Microscopy Sciences

Supplier: Electron Microscopy Sciences
Nunc™ Thermanox™
72273 72275-01 72286 72274-01 72285 72272 72270 72275-05 72282 72271 72281 72283 72274-05 72280
100500-870BX 717.32 USD InStock
100500-870 100500-872 100500-874 100500-864 100500-876 100500-866 100502-644 100500-880 100501-064 100501-062 100500-878 100500-868 100501-068 100501-066
Thermanox® Cover Slips, Electron Microscopy Sciences
Cover Glasses
NUNC™ Brand Thernanox®, or TMX coverslips are made from a polymer (in the polyolefin family) that is highly resistant to most chemicals.

  • Flat and remain flat
  • 0.2mm (No. 2) thickness
  • Resistant to commonly used solvents
  • Safe and easy to handle
  • Auto fluorescent in the range 380 to 545 nm
  • Thernanox® cover slips can be autoclaved at 20 minutes steam, 20 minutes dry at 120°C

Thermanox® plastic is resistant to alcohols, aldehydes, hydrocarbons, dilute acids (>10%) and dilute alkalis (>2%). TMX has limited resistance to chlorinated hydro-carbons, however, it is not resistant to concentrated acids or bases.Thermanox® plastic is a flexible, trans-parent polymer that can be sectioned using a microtome and is able to withstand high temp-eratures (temperature range –70°C to +150°C). Thermanox® coverslips are culture treated on one side for enhanced cell attachment and growth. The treated side is packaged face up toward the label.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty