Would you like to log in again?
Premium Abrasive Cut-off Wheels for Sectioning, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
50250-01 50250-06 50255-01 50250-03 50250-08 50250-09 50255-04 50255-08 50250-10 50255-10 50255-07 50250-05 50250-04 50250-07 50255-05 50255-06 50255-03 50255-09
100500-940BX 73.08 USD InStock
100500-940 100500-930 100500-920 100500-910 100500-932 100502-650 100502-648 100500-922 100500-912 100500-934 100500-924 100500-936 100500-914 100500-926 100500-916 100500-938 100500-928 100500-918
Premium Abrasive Cut-off Wheels for Sectioning, Electron Microscopy Sciences
Knives
Silicone Carbide disc are the finest wet or dry abrasive discs for sample preparation
Wheels are available in 9 different grit sizes, ranging from coarse to fine, on plain back or water resistant pressure sensitive adhesive back.
Pressure sensitive adhesive backed discs eliminates the need to transfer to plates. Silicon Carbide Abrasive Cut-Off Wheels offered long life, and affordably priced sectioning. All wheels are 9" (228.6 mm) diameter 0.062” (1.6 mm) thick; 1¼” (31mm) arbor.
Ordering information: Available in 8" diameter discs as a standard and 12" discs available upon request.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty