Would you like to log in again?
Polysine™ Microscope Adhesion Slide, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
63412-02 63412-01
100501-006BX 204.4 USD InStock
100501-006 100501-004
Polysine™ Microscope Adhesion Slide, Electron Microscopy Sciences
Slides Adhesive Slides
The permanent bio-adhesive material on our Polysine™ slide attracts frozen and paraffin embedded sections, centrifuge preparation and cytology smears.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty