Would you like to log in again?
4 oz. Specimen Container, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
64231-20
100501-144BG 19.86 USD InStock
100501-144
4 oz. Specimen Container, Electron Microscopy Sciences
Sample Containers General Purpose Containers
This 4 oz. specimen container provides a “full-turn” screw thread cap, which offers an excellent “leak-proof” seal that meets the NCCLS Specimen Handling Guidelines for primary containers.
These versatile, graduated, polypropylene containers with polyethylene caps can be used for almost any liquid or solid specimen for either storage or transport purposes. They come with color coded caps: green for individually wrapped sterile caps, white for non-sterile ones.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty