Would you like to log in again?
Anemometer, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
72082
100501-344EA 398.92 USD InStock
100501-344
Anemometer, Electron Microscopy Sciences
Anemometers
Perfect for use in fume hoods, environmental studies, or for air flow check. A highly accurate, hand-held unit, only 3 ozs. Allows you to select readout in knots, feet per minute, meters per second, or miles per hour. To insure maximum sensitivity and accuracy, the turbine is suspended on saphire jewel bearings, and its rotation is sensed by an infrared light beam, which adds no friction. The resulting signal is pro-cessed electrically by an LSI (Large Scale Integrated) circuit. Measurements: 4.5 x 2.5 x 1.5".
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty